Learn More
3-Nitro-L-tyrosine, TRC
Product Code. 17959984
Change view
Click to view available options
Quantity:
25 mg
1 g
5 g
Unit Size:
1g
25mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17959984 25 mg 25mg
17907534 5 g 5g
17957254 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17959984 Supplier Toronto Research Chemicals Supplier No. TRCN59700025MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 621-44-3
Molecular Formula C9 H10 N2 O5
Molecular Weight (g/mol) 226.19
Synonym L-Tyrosine, 3-nitro- (9CI, ACI), 3-Nitro-L-tyrosine (ACI), Tyrosine, 3-nitro-, L- (8CI), (2S)-2-Amino-3-(4-hydroxy-3-nitrophenyl)propanoic acid, (2S)-2-Azaniumyl-3-(4-hydroxy-3-nitrophenyl)propanoate, Nitrotyrosine, NSC 37413
IUPAC Name (2S)-2-amino-3-(4-hydroxy-3-nitrophenyl)propanoic acid
SMILES N[C@@H](Cc1ccc(O)c(c1)[N+](=O)[O-])C(=O)O
Specifications
InChI Formula InChI=1S/C9H10N2O5/c10-6(9(13)14)3-5-1-2-8(12)7(4-5)11(15)16/h1-2,4,6,12H,3,10H2,(H,13,14)/t6-/m0/s1
Quantity 25 mg
Formula Weight 226.059
Shipping Condition Room Temperature
Chemical Name or Material 3-Nitro-L-tyrosine