Learn More
3-Nitro-N-phenylbenzamide, 97%, Thermo Scientific™
Product Code. 15497178
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15497178 250 mg 250mg
15487178 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15497178 Supplier Thermo Scientific Alfa Aesar Supplier No. H60695.MD
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2243-73-4
Molecular Formula C13H10N2O3
Molecular Weight (g/mol) 242.234
MDL Number MFCD00017021
InChI Key VWIRWLAPFZXYSL-UHFFFAOYSA-N
Synonym 3-nitrobenzanilide, 3-nitro-n-phenyl-benzamide, benzamide, 3-nitro-n-phenyl, 3-nitrophenyl-n-benzamide, 3-nitro-n-phenyl benzamide
PubChem CID 347755
IUPAC Name 3-nitro-N-phenylbenzamide
SMILES C1=CC=C(C=C1)NC(=O)C2=CC(=CC=C2)[N+](=O)[O-]
Specifications
Quantity 250 mg
Formula Weight 242.23
Percent Purity 97%
Chemical Name or Material 3-Nitro-N-phenylbenzamide