missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrophthalonitrile, 99%

Product Code. 11470828
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11470828 5 g 5g
11480828 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11470828 Supplier Thermo Scientific Alfa Aesar Supplier No. B25269.06
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 51762-67-5 | C8H3N3O2 | 173.13 g/mol

3-Nitrophthalonitrile is a useful dye for biological research purposes. It is used as an intermediate in the manufacture of pharmaceuticals, chemicals, dyestuff and phthalocyanine blue pigments.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Nitrophthalonitrile is a useful dye for biological research purposes. It is used as an intermediate in the manufacture of pharmaceuticals, chemicals, dyestuff and phthalocyanine blue pigments.

Solubility
Soluble in methanol. Insoluble in water.

Notes
Incombatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51762-67-5
Molecular Formula C8H3N3O2
Molecular Weight (g/mol) 173.13
MDL Number MFCD00191558
InChI Key UZJZIZFCQFZDHP-UHFFFAOYSA-N
Synonym 3-nitrophthalonitrile, 2,3-dicyanonitrobenzene, 1,2-benzenedicarbonitrile, 3-nitro, 3-nitrophthalodinitrile, 1,2-dicyano-3-nitrobenzene, 3-nitro-1,2-benzenedicarbonitrile, nitrophthalonitrile, 3-nitro-phthalonitrile, 3-nitrophthalonitrile?, acmc-209kvo
PubChem CID 162652
IUPAC Name 3-nitrobenzene-1,2-dicarbonitrile
SMILES [O-][N+](=O)C1=CC=CC(C#N)=C1C#N

Specifications

Melting Point 162°C to 165°C
Quantity 5 g
Beilstein 2263686
Solubility Information Soluble in methanol. Insoluble in water.
Formula Weight 173.13
Percent Purity 99%
Chemical Name or Material 3-Nitrophthalonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.