missing translation for 'onlineSavingsMsg'
Learn More

3-(o-Hydroxyphenyl)-5-phenyl-2-cyclohexen-1-one, TRC

Product Code. 30399034
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30399034 100 mg
30392447 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30399034 Supplier Toronto Research Chemicals Supplier No. TRCH949435100MG
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15156-56-6
Molecular Formula C18 H16 O2
Molecular Weight (g/mol) 264.32
Synonym 2-Cyclohexen-1-one, 3-(2-hydroxyphenyl)-5-phenyl-, 2-Cyclohexen-1-one, 3-(o-hydroxyphenyl)-5-phenyl- (8CI), 3-(2-Hydroxyphenyl)-5-phenyl-2-cyclohexen-1-one, (5RS)-3-(2-Hydroxyphenyl)-5-phenylcyclohex-2-enone, 3-(o-hydroxyphenyl)-5-phenyl-2-cyclohexen-1-one
IUPAC Name 3-(2-hydroxyphenyl)-5-phenylcyclohex-2-en-1-one
SMILES Oc1ccccc1C2=CC(=O)CC(C2)c3ccccc3

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C18H16O2/c19-16-11-14(13-6-2-1-3-7-13)10-15(12-16)17-8-4-5-9-18(17)20/h1-9,12,14,20H,10-11H2
Formula Weight 264.115
Percent Purity >95%
Chemical Name or Material (5RS)-3-(2-Hydroxyphenyl)-5-phenylcyclohex-2-enone
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.