missing translation for 'onlineSavingsMsg'
Learn More

3-O-Methyl Carbidopa, TRC

Product Code. 30007299
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30007299 100 mg 100mg
30003541 50 mg 50mg
30005721 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007299 Supplier Toronto Research Chemicals Supplier No. TRCM294590100MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85933-19-3
Molecular Formula C11 H16 N2 O4
Molecular Weight (g/mol) 240.26
Synonym α-Hydrazinyl-4-hydroxy-3-methoxy-α-methyl-benzenepropanoic acid, Benzenepropanoic acid, α-hydrazino-4-hydroxy-3-methoxy-α-methyl- (9CI), Hydrocinnamic acid, α-hydrazino-4-hydroxy-3-methoxy-α-methyl- (7CI)
IUPAC Name 2-hydrazinyl-3-(4-hydroxy-3-methoxyphenyl)-2-methylpropanoic acid
SMILES COc1cc(CC(C)(NN)C(=O)O)ccc1O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C11H16N2O4/c1-11(13-12,10(15)16)6-7-3-4-8(14)9(5-7)17-2/h3-5,13-14H,6,12H2,1-2H3,(H,15,16)
Formula Weight 240.111
Chemical Name or Material 3-O-Methylcarbidopa
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.