missing translation for 'onlineSavingsMsg'
Learn More

3-O-Methyl Quercetin, TRC

Product Code. 30006658
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006658 1 mg 1mg
30003686 5 mg 5mg
30003643 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30006658 Supplier Toronto Research Chemicals Supplier No. TRCM3265451MG
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1486-70-0
Molecular Formula C16 H12 O7
Molecular Weight (g/mol) 316.26
Synonym 2-(3,4-Dihydroxyphenyl)-5,7-dihydroxy-3-methoxy-4H-1-benzopyran-4-one, 3-Methoxy-3',4',5,7-tetrahydroxyflavone, 3-Methylquercetin, 3-Methylquercetol, 3',4',5,7-Tetrahydroxy-3-methoxyflavone, 5,7,3',4'-Tetrahydroxy-3-methoxyflavone, NSC 154016, Quercetin 3-Methyl Ether, Quercetin-3-O-methyl Ether;
IUPAC Name 2-(3,4-dihydroxyphenyl)-5,7-dihydroxy-3-methoxychromen-4-one
SMILES COC1=C(Oc2cc(O)cc(O)c2C1=O)c3ccc(O)c(O)c3

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C16H12O7/c1-22-16-14(21)13-11(20)5-8(17)6-12(13)23-15(16)7-2-3-9(18)10(19)4-7/h2-6,17-20H,1H3
Formula Weight 316.0583
Chemical Name or Material 3',4',5,7-Tetrahydroxy-3-methoxyflavone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.