missing translation for 'onlineSavingsMsg'
Learn More

3-Pentanol, TRC

Product Code. 30136957
Change view
Click to view available options
Quantity:
50 mL
100 mL
500 mL
Unit Size:
100mL
500mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136957 500 mL 500mL
30136236 50 mL 50mL
17974235 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136957 Supplier Toronto Research Chemicals Supplier No. TRCP227295500ML
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 584-02-1
Molecular Formula C5 H12 O
Molecular Weight (g/mol) 88.15
Synonym 3-Pentanol (8CI, 9CI, ACI), 1-Ethyl-1-propanol, 1-Ethylpropyl alcohol, 3-Hydroxypentane, 3-Pentyl alcohol, Diethyl carbinol, NSC 8654, sec-Amyl alcohol, sec-Pentanol, sec-Pentyl alcohol
IUPAC Name pentan-3-ol
SMILES CCC(O)CC

Specifications

Quantity 500 mL
InChI Formula InChI=1S/C5H12O/c1-3-5(6)4-2/h5-6H,3-4H2,1-2H3
Formula Weight 88.0888
Chemical Name or Material 3-Pentanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.