missing translation for 'onlineSavingsMsg'
Learn More

3-Phenyl-2-oxazolidinone, TRC

Product Code. 30428749
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30428749 100 mg -
30428113 50 mg 50mg
30428232 500 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30428749 Supplier Toronto Research Chemicals Supplier No. TRCP336023100MG
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 703-56-0
Molecular Formula C9H9NO2
Molecular Weight (g/mol) 163.17
IUPAC Name 3-phenyl-1,3-oxazolidin-2-one
SMILES O=C1OCCN1c2ccccc2

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C9H9NO2/c11-9-10(6-7-12-9)8-4-2-1-3-5-8/h1-5H,6-7H2
Formula Weight 163.0633
Chemical Name or Material 3-Phenyl-2-oxazolidinone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.