Learn More
3-Phenyl-2-propenoic acid benzyl ester, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 103-41-3 |
| Molecular Formula | C16H14O2 |
| MDL Number | MFCD00004789 |
| Quantity | 100 g |
| SMILES | O=C(OCC1=CC=CC=C1)/C=C/C2=CC=CC=C2 |
| Molecular Weight (g/mol) | 238.28 |
| Percent Purity | 98% |
| Chemical Name or Material | 3-Phenyl-2-propenoic acid benzyl ester |