Learn More
3-Phenyl-2-propyn-1-ol, 98+%
Description
3-Phenyl-2-propyn-1-ol is an phenyl acetylene derivative with inhibitory effect on nitrification in soil.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Density | 1.078 g/mL |
| Boiling Point | 129°C to 130°C (10 mmHg) |
| Flash Point | >110°C (230°F) |
| Refractive Index | 1.5867 |
| Quantity | 10 g |
| Beilstein | 2040911 |
| Solubility Information | Soluble in ether,acetone,benzene,chloroform,ethyl Acetate,methanol. |
| Formula Weight | 132.16 |
| Percent Purity | ≥98% |
| Physical Form | Liquid |
| Show More |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.