missing translation for 'onlineSavingsMsg'
Learn More

3-(Piperazin-1-yl)-1,2-benzisoxazole (>90%), TRC

Product Code. 30426286
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30426286 100 mg -
30410428 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30426286 Supplier Toronto Research Chemicals Supplier No. TRCP475523100MG
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87691-89-2
Molecular Formula C11H13N3O
Molecular Weight (g/mol) 203.24
Synonym 1-(Benzisoxazol-3-yl)piperazine, 1-(Benzo[d]isoxazol-3-yl)piperazine, 3-(Piperazin-1-yl)benzo[d]isoxazole, 3-Piperazin-1-yl-1,2-benzoxazole, 3-Piperazino-1,2-benzisoxazole
IUPAC Name 3-piperazin-1-yl-1,2-benzoxazole
SMILES C1CN(CCN1)c2noc3ccccc23

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C11H13N3O/c1-2-4-10-9(3-1)11(13-15-10)14-7-5-12-6-8-14/h1-4,12H,5-8H2
Formula Weight 203.1059
Chemical Name or Material 3-(Piperazin-1-yl)-1,2-benzisoxazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.