missing translation for 'onlineSavingsMsg'
Learn More

3-Pyridin-3-ylaniline, 97%, Thermo Scientific™

Product Code. 10249722
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10249722 1 g Amber Glass Bottle 1g
10239722 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10249722 Supplier Thermo Scientific Maybridge Supplier No. CC39814DA
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 57976-57-5
Molecular Formula C11H10N2
Molecular Weight (g/mol) 170.215
InChI Key YTJQJGKMRLQBJP-UHFFFAOYSA-N
Synonym 3-pyridin-3-yl benzenamine, 3-pyridin-3-yl aniline, 3-3-aminophenyl pyridine, 3-3-pyridyl aniline, benzenamine, 3-3-pyridinyl, 3-3-pyridinyl aniline, 3-pyridin-3-yl anilin, 3-3-pyridyl phenylamine, 3-pyridine-3-yl aniline, 3-pyridin-3-yl-phenylamine
PubChem CID 459521
IUPAC Name 3-pyridin-3-ylaniline
SMILES C1=CC(=CC(=C1)N)C2=CN=CC=C2

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 170.21
Percent Purity 97%
Chemical Name or Material 3-Pyridin-3-ylaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.