missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethoxy)benzenesulfonyl chloride, 95%, Thermo Scientific™

Product Code. 11358946
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11358946 250 mg 250mg
11338946 1 g 1g
11348946 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11358946 Supplier Thermo Scientific Alfa Aesar Supplier No. L19552.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 220227-84-9
Molecular Formula C7H4ClF3O3S
Molecular Weight (g/mol) 260.611
MDL Number MFCD01091016
InChI Key DODDSXTWDSJCDN-UHFFFAOYSA-N
Synonym 3-trifluoromethoxy benzenesulfonyl chloride, 3-trifluoromethoxy benzene-1-sulfonyl chloride, 3-trifluoromethoxy benzenesulphonyl chloride, 3-trifluoromethoxy-benzenesulfonyl chloride, 3-trifluoromethoxybenzenesulfonylchloride, 3-trifluoromethyl oxy benzenesulfonyl chloride, benzenesulfonyl chloride, 3-trifluoromethoxy, pubchem5187, acmc-1ce4e
PubChem CID 2777218
IUPAC Name 3-(trifluoromethoxy)benzenesulfonyl chloride
SMILES C1=CC(=CC(=C1)S(=O)(=O)Cl)OC(F)(F)F

Specifications

Melting Point 229°C to 230°C
Density 1.53
Boiling Point 73°C to 74°C (0.7 mmHg)
Refractive Index 1.4754
Quantity 250 mg
UN Number UN3265
Sensitivity Moisture sensitive
Formula Weight 260.62
Percent Purity 95%
Chemical Name or Material 3-(Trifluoromethoxy)benzenesulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.