missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)-1H-pyrazole-5-carboxylic Acid Potassium, TRC

Product Code. 30453228
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30453228 500 mg 500mg
30456655 100 mg 100mg
30451428 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30453228 Supplier Toronto Research Chemicals Supplier No. TRCT897605500MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-(Trifluoromethyl)-1H-pyrazole-5-carboxylic Acid Potassium
Analyte or Component Names 3-(Trifluoromethyl)-1H-pyrazole-5-carboxylic Acid Potassium
Shipping Condition Room Temperature
Molecular Formula C5 H3 N2 O2 F3 .xK
InChI Formula InChI=1S/C5H3F3N2O2.K/c6-5(7,8)3-1-2(4(11)12)9-10-3;/h1H,(H,9,10)(H,11,12);
Solution Type Neat
SMILES O=C(O)C1=CC(C(F)(F)F)=NN1.[xK]
IUPAC Name 3-(trifluoromethyl)-1H-pyrazole-5-carboxylic acid potassium
Industry Type Pharmaceutical
Molecular Weight (g/mol) 219.18
Quantity 500 mg
Formula Weight 218.9784
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.