missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)benzaldehyde, 97%

Product Code. 10062570 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10062570 25 g Glass bottle 25g
10206440 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10062570 Supplier Thermo Scientific Acros Supplier No. 188440250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 454-89-7 | C8H5F3O | 174.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 454-89-7
Molecular Formula C8H5F3O
Molecular Weight (g/mol) 174.12
MDL Number MFCD00003373
InChI Key NMTUHPSKJJYGML-UHFFFAOYSA-N
Synonym 3-trifluoromethyl benzaldehyde, m-trifluoromethylbenzaldehyde, benzaldehyde, 3-trifluoromethyl, alpha,alpha,alpha-trifluoro-m-tolualdehyde, alpha,alpha,alpha-trifluoro-3-tolualdehyde, unii-at7h4h5u3e, a,a,a-trifluoro-m-tolualdehyde, 3-trifluoromethyl-benzaldehyde, mtf-bad, 3-formylbenzotrifluoride
PubChem CID 67990
IUPAC Name 3-(trifluoromethyl)benzaldehyde
SMILES C1=CC(=CC(=C1)C(F)(F)F)C=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Orange
Density 1.3000g/mL
Boiling Point 83°C to 86°C (30.0 mmHg)
Flash Point 68°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CF3C6H4CHO
Refractive Index 1.4639 to 1.4659
Quantity 25 g
Beilstein 07, III, 1016
Specific Gravity 1.3
Formula Weight 174.12
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-(Trifluoromethyl)benzaldehyde, 97%
Show More Show Less
compliance-icons
Product Identifier
  • a,a,a-Trifluoro-m-tolualdehyde
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.