missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)benzene-1-sulfonyl chloride, 97%, Maybridge

Product Code. 10103454
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10103454 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10103454 Supplier Thermo Scientific Maybridge Supplier No. BTB06459EA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 777-44-6
Assay Percent Range 97%
Molecular Formula C7H4ClF3O2S
Quantity 10 g
Synonym 3-trifluoromethyl benzenesulfonyl chloride, 3-trifluoromethyl benzene-1-sulfonyl chloride, m-trifluoromethylbenzenesulfonyl chloride, 3-trifluoromethyl benzenesulphonyl chloride, 3-trifluoromethylbenzenesulfochloride, 3-trifluoromethyl benzenesulfonylchloride, benzenesulfonyl chloride, 3-trifluoromethyl, 3-chlorosulphonyl benzotrifluoride, m-trifluoromethylphenylsulfonyl chloride, 3-trifluoromethyl-benzenesulfonyl chloride
InChI Key ONCAZCNPWWQQMW-UHFFFAOYSA-N
SMILES C1=CC(=CC(=C1)S(=O)(=O)Cl)C(F)(F)F
IUPAC Name 3-(trifluoromethyl)benzenesulfonyl chloride
Molecular Weight (g/mol) 244.612
PubChem CID 2733250
Formula Weight 244.62
Chemical Name or Material 3-(Trifluoromethyl)benzene-1-sulfonyl chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.