missing translation for 'onlineSavingsMsg'
Learn More

(3-(Trifluoromethyl)phenyl)boronic acid, 98%, BLD Pharm™

Product Code. 31089930
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
100 g
500 g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31089930 500 g
31080365 1 g
31089815 5 g
31084402 100 g
31082438 25 g
31077539 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 31089930 Supplier BLD Pharm Supplier No. BD10398500g
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 1423-26-3
Molecular Formula C7H6BF3O2
MDL Number MFCD00151854
Quantity 500 g
SMILES FC(C1=CC(B(O)O)=CC=C1)(F)F
Molecular Weight (g/mol) 189.93
Percent Purity 98%
Chemical Name or Material (3-(Trifluoromethyl)phenyl)boronic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.