missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)phenyl isothiocyanate, 98%

Product Code. 11352547
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11352547 1 g 1g
11362547 5 g 5g
11372547 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11352547 Supplier Thermo Scientific Alfa Aesar Supplier No. L11646.03
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 1840-19-3 | C8H4F3NS | 203.182 g/mol

Used as pharamaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as pharamaceutical intermediate.

Solubility
Hydrolyzes in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1840-19-3
Molecular Formula C8H4F3NS
Molecular Weight (g/mol) 203.182
MDL Number MFCD00004806
InChI Key GFEPANUKFYVALF-UHFFFAOYSA-N
Synonym 3-trifluoromethyl phenyl isothiocyanate, 1-isothiocyanato-3-trifluoromethyl benzene, 3-trifluoromethyl phenylisothiocyanate, benzene, 1-isothiocyanato-3-trifluoromethyl, m-trifluoromethyl phenyl isothiocyanate, alpha,alpha,alpha-trifluoro-m-tolyl isothiocyanate, 3-trifluoromethylphenyl isothiocyanate, 1-isothiocyanato-3-trifluoromethyl-benzene, benzene,1-isothiocyanato-3-trifluoromethyl
PubChem CID 137232
IUPAC Name 1-isothiocyanato-3-(trifluoromethyl)benzene
SMILES C1=CC(=CC(=C1)N=C=S)C(F)(F)F

Specifications

Density 1.335
Boiling Point 208°C
Flash Point 95°C (203°F)
Odor Pungent
Refractive Index 1.5547
Quantity 1 g
UN Number UN2922
Beilstein 781205
Sensitivity Moisture sensitive
Solubility Information Hydrolyzes in water.
Formula Weight 203.19
Percent Purity 98%
Chemical Name or Material 3-(Trifluoromethyl)phenyl isothiocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.