missing translation for 'onlineSavingsMsg'
Learn More

[3-(Trifluoromethyl)phenyl]methanethiol, Tech., Thermo Scientific™

Product Code. 10643613
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10643613 10 g 10g
10325012 1 g 1g
11396955 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10643613 Supplier Thermo Scientific Maybridge Supplier No. TL00142EA
 Restricted Item
Estimated Shipment: 06-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 25697-55-6
Molecular Formula C8H7F3S
Molecular Weight (g/mol) 192.199
MDL Number MFCD00042435
InChI Key CQIQWIMXCPTQPJ-UHFFFAOYSA-N
Synonym 3-trifluoromethyl benzyl mercaptan, 3-trifluoromethyl phenyl methanethiol, 3-trifluoromethylbenzyl mercaptan, 3-trifluoromethyl phenylmethanethiol, 3-trifluoromethyl benzylthiol, m-trifluoromethylbenzyl mercaptan, 3-trifluoromethyl benzylmercaptan, 3-trifluoromethyl phenyl methane-1-thiol, acmc-1cnzu
PubChem CID 141236
IUPAC Name [3-(trifluoromethyl)phenyl]methanethiol
SMILES C1=CC(=CC(=C1)C(F)(F)F)CS

Specifications

Packaging Amber Glass Bottle
Linear Formula CF3C6H4CH2SH
Quantity 10 g
Formula Weight 192.2
Percent Purity Tech.
Grade Technical
Chemical Name or Material [3-(Trifluoromethyl)phenyl]methanethiol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.