missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)thiobenzamide, 97%

Product Code. 15433447
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15433447 250 mg 250mg
11481170 1 g 1g
11491170 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15433447 Supplier Thermo Scientific Alfa Aesar Supplier No. H26311.MD
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 53515-17-6 | C8H6F3NS | 205.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53515-17-6
Molecular Formula C8H6F3NS
Molecular Weight (g/mol) 205.20
MDL Number MFCD01567258
InChI Key WOJQBHSZMFRQIR-UHFFFAOYSA-N
Synonym 3-trifluoromethyl thiobenzamide, 3-trifluoromethylthiobenzamide, 3-trifluoromethyl benzenecarbothioamide, 3-trifluoromethyl benzene-1-carbothioamide, 3-trifluoromethyl-thiobenzamide, amino 3-trifluoromethyl phenyl methane-1-thione, acmc-20amye, pubchem5483, maybridge1_008312
PubChem CID 2777849
IUPAC Name 3-(trifluoromethyl)benzenecarbothioamide
SMILES NC(=S)C1=CC(=CC=C1)C(F)(F)F

Specifications

Quantity 250 mg
UN Number UN2811
Solubility Information Insoluble in water.
Formula Weight 205.2
Percent Purity 97%
Chemical Name or Material 3-(Trifluoromethyl)thiobenzamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.