Learn More
3-Trimethylsilyl-1-propylboronic acid pinacol ester, 94%, Thermo Scientific™
Product Code. 15437745
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Product Code. 15437745
Brand: Thermo Scientific Alfa Aesar H28413.MD
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 165904-32-5
Molecular Formula C12H27BO2Si
Molecular Weight (g/mol) 242.241
MDL Number MFCD09953502
InChI Key SVYVMKANVAUFQP-UHFFFAOYSA-N
Synonym 3-trimethylsilyl-1-propylboronic acid pinacol ester, 2-3-trimethylsilyl propyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-3-trimethylsilylpropyl-1,3,2-dioxoborolane, trimethyl 3-tetramethyl-1,3,2-dioxaborolan-2-yl propyl silane, trimethyl 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl propyl silane
PubChem CID 11032057
IUPAC Name trimethyl-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)propyl]silane
SMILES B1(OC(C(O1)(C)C)(C)C)CCC[Si](C)(C)C
Specifications
Quantity 250 mg
Formula Weight 242.24
Percent Purity 94%
Chemical Name or Material 3-Trimethylsilyl-1-propylboronic acid pinacol ester