missing translation for 'onlineSavingsMsg'
Learn More

3-Trimethylsilyl-2-propyn-1-ol, 99%, Thermo Scientific™

Product Code. 10720961 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10720961 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10720961 Supplier Thermo Scientific Acros Supplier No. 313890010
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 5272-36-6
CAS Min % 98.5
CAS Max % 100.0
Color Yellow
Density 0.8650g/mL
Boiling Point 76°C (20.0 mmHg)
Flash Point 66°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C6H12OSi
Linear Formula (CH3)3SIC∽CCH2OH
Refractive Index 1.4500 to 1.452
MDL Number MFCD00002913
Quantity 1 g
Specific Gravity 0.865
Synonym 3-trimethylsilyl-2-propyn-1-ol, 3-trimethylsilyl propargyl alcohol, 2-propyn-1-ol, 3-trimethylsilyl, 3-trimethylsilyl prop-2-yn-1-ol, 1-trimethylsilylpropargyl alcohol, acmc-209l1b, 3-trimethylsilylpropargyl alcohol, 3-trimethylsilanyl-prop-2-yn-1-ol, 3-trimethylsilyl-2-propyn-1-ol #
InChI Key ZVGCJDPEKKEYES-UHFFFAOYSA-N
SMILES C[Si](C)(C)C#CCO
IUPAC Name 3-trimethylsilylprop-2-yn-1-ol
Molecular Weight (g/mol) 128.25
PubChem CID 78930
Formula Weight 128.25
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 3-Trimethylsilyl-2-propyn-1-ol, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.