missing translation for 'onlineSavingsMsg'
Learn More

3-Vinylbenzoic acid, 96%

Product Code. 15415297
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15415297 5 g 5g
15405297 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15415297 Supplier Thermo Scientific Alfa Aesar Supplier No. H32556.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 28447-20-3 | C9H8O2 | 148.161 g/mol

Polymers were created by the imprinting of tri-O-acetyladenosine in acrylic polymer matrix with the use of methacrylic acid (MAA) or 3-vinylbenzoic acid (VBA) as a functional monomer and ethylene glycol dimethacrylate as a cross-linking agent. Nine active ester monomers based on 4-vinylbenzoic acid have been synthesized. Under free radical polymerization conditions these monomers could successfully be polymerized yielding reactive polymers

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Polymers were created by the imprinting of tri-O-acetyladenosine in acrylic polymer matrix with the use of methacrylic acid (MAA) or 3-vinylbenzoic acid (VBA) as a functional monomer and ethylene glycol dimethacrylate as a cross-linking agent. Nine active ester monomers based on 4-vinylbenzoic acid have been synthesized. Under free radical polymerization conditions these monomers could successfully be polymerized yielding reactive polymers

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat, light, ultraviolet radiations, free radical initiators.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28447-20-3
Molecular Formula C9H8O2
Molecular Weight (g/mol) 148.161
MDL Number MFCD00157038
InChI Key VWXZFDWVWMQRQR-UHFFFAOYSA-N
PubChem CID 4438231
IUPAC Name 3-ethenylbenzoic acid
SMILES C=CC1=CC(=CC=C1)C(=O)O

Specifications

Melting Point 95°C
Quantity 5 g
Synonym 3-vinylbenzoic acid, 3-vinyl benzoic acid, 3-ethenyl-benzoic acid, benzoic acid,3-ethenyl, benzoic acid, 3-ethenyl, benzoic acid, 3-ethenyl-9ci
Formula Weight 148.16
Percent Purity 96%
Chemical Name or Material 3-Vinylbenzoic acid
compliance-icons
Product Identifier
  • 3-Vinylbenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 3-Vinylbenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.