missing translation for 'onlineSavingsMsg'
Learn More

3(Z)-Nonenol, TRC

Product Code. 30483560
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. Quantity unitSize
30483560 1 g 1g
1 options
This item is not returnable. View return policy

Product Code. 30483560

Brand: Toronto Research Chemicals TRCN6495601G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3(Z)-Nonenol
CAS 10340-23-5
Molecular Formula C9H18O
Quantity 1 g
Synonym (Z)-3-Nonen-1-ol, (Z)-3-Nonen-1ol, (Z)-3-Nonen-1-ol, 3(Z)-Nonenol, cis-3-Nonen-1-ol, cis-3Nonenol
InChI Formula InChI=1S/C9H18O/c1-2-3-4-5-6-7-8-9-10/h6-7,10H,2-5,8-9H2,1H3/b7-6-
SMILES CCCCC\C=C/CCO
IUPAC Name (Z)-non-3-en-1-ol
Molecular Weight (g/mol) 142.24
Formula Weight 142.1358
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.