Learn More
Sampling Systems™ 304 Stainless Steel Bowls
Product Code. 15839691 Shop All Sampling Systems Products
Change view
Click to view available options
Capacity (Metric):
1000 mL
2 L
4 L
Length (Metric):
170 mm
220 mm
270 mm
Height (Metric):
85 mm
110 mm
130 mm
Width (Metric):
170 mm
220 mm
270 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Length (Metric) Height (Metric) Width (Metric) unitSize
15839691 4 L 270 mm 130 mm 270 mm Each
15829691 2 L 220 mm 110 mm 220 mm Each
15819691 1000 mL 170 mm 85 mm 170 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15839691 Supplier Sampling Systems™ Supplier No. A5344000
Only null left
Add to basket
Add to basket
Sampling Systems™ 304 Stainless Steel Bowls are ideal for hygienic applications.
  • 304 Stainless Steel
  • Crevice free design
  • Polished finish
  • Range of sizes
Specifications
Product Type Bowl
Material Stainless Steel (AISI 304)
Capacity (Metric) 4 L
Height (Metric) 130 mm
Length (Metric) 270 mm
Width (Metric) 270 mm