missing translation for 'onlineSavingsMsg'
Learn More

(2R,4R)-rel-1,2,3,4-Tetrahydro-6-methyl-2,4-bis(2-nitrophenyl)-5-pyrimidinecarboxylic Acid Methyl Ester, TRC

Product Code. 30421142
Change view
Click to view available options
Quantity:
100 mg
250 mg
2.5 g
Unit Size:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30421142 2.5 g 2.5g
30412684 250 mg -
30424504 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30421142 Supplier Toronto Research Chemicals Supplier No. TRCT2956002.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80742-11-6
Molecular Formula C19 H18 N4 O6
Molecular Weight (g/mol) 398.37
Synonym 5-Pyrimidinecarboxylic acid, 1,2,3,4-tetrahydro-6-methyl-2,4-bis(2-nitrophenyl)-, methyl ester, (2R,4R)-rel-, 5-Pyrimidinecarboxylic acid, 1,2,3,4-tetrahydro-6-methyl-2,4-bis(2-nitrophenyl)-, methyl ester, cis-(±)-, 5-Pyrimidinecarboxylic acid, 1,2,3,4-tetrahydro-6-methyl-2,4-bis(2-nitrophenyl)-, methyl ester, cis-, Methyl (2RS,4RS)-6-Methyl-2,4-bis(2-nitrophenyl)-1,2,3,4-tetrahydropyrimidine-5-carboxylate
IUPAC Name methyl (2R,4R)-6-methyl-2,4-bis(2-nitrophenyl)-1,2,3,4-tetrahydropyrimidine-5-carboxylate
SMILES COC(=O)C1=C(C)N[C@@H](N[C@@H]1c2ccccc2[N+](=O)[O-])c3ccccc3[N+](=O)[O-]

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C19H18N4O6/c1-11-16(19(24)29-2)17(12-7-3-5-9-14(12)22(25)26)21-18(20-11)13-8-4-6-10-15(13)23(27)28/h3-10,17-18,20-21H,1-2H3/t17-,18+/m1/s1
Formula Weight 398.1226
Chemical Name or Material (2R,4R)-rel-1,2,3,4-Tetrahydro-6-methyl-2,4-bis(2-nitrophenyl)-5-pyrimidinecarboxylic acid methyl ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.