Learn More
Consort 3050 Bio-Electric Analyzer
Product Code. 11905458
Change view
Click to view available options
Range (pH):
-2 to 16
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Range (pH) unitSize
11905458 -2 to 16 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Product Code. 11905458 Supplier Consort Supplier No. C3050
Only null left
Add to basket
Add to basket
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Capable of measuring according to Vincent’s method. Consort 3050 Bio-Electric Analyzer is a three-channel instrument that can measure and display all parameters at once using electrodes connected by gold-plated BNC connections, plus two temperature inputs via banana connectors.
This instrument is built with the same quality as the instruments in the whole C3000 series. As such, it has the same features as the other versions.
  • Multichannel up to 3 measurements can be performed at the same time (2X / sec. / channel)
  • Parameters according to Vincent method can be shown on the display simultaneously or individually
  • No interference between pH/ORP/ion and conductivity electrodes in the same solution
  • Pre-programmed pH buffers and EC standards, plus add up to 5 pH buffers and 3 EC standards
  • Custom calibration tables provide complete buffer/standard-temperature relation information
  • Direct rH2 and μW measurement when using a pH, ORP and EC electrode
  • Wide Display shows all channels at the same time including temperature and date/time
  • Stability indicator ensures visualization when the measurement has stabilized
  • Stability algorithm ensures stable readings with ability to detect fast changes
  • Hold function allows to freeze the display
  • Selectable resolution for more stable readings on mV and pH
  • Range lock for conductivity measurements
  • Capacitive compensation eliminates electrode/cable capacitive component with low conductivities
  • Galvanic isolated USB interface eliminates ground loop effects when connected to a PC
  • Pre-programmed standards
    pH: 1.68, 2.00, 4.00, 4.01, 6.87, 6.99, 9.18, 9.21, 10.01, 12.00, 12.45 (at 25°C)
    Conductivity: 1413 μS/cm, 12.88 mS/cm, 111.8 mS/cm (at 25°C)
  • Free software and firmware updates downloadable from www.consort.be
Specifications
Accuracy (Temperature) 0.1°C
Accuracy (mV) 0.1% ± 1 digit
Data Management Details Manual or Timed, Interval: 1 to 9999 sec.
Outputs USB, RS-232
Calibration GLP, Reminder: 0 to 999 h
Range (pH) -2 to 16
Resolution (pH) 0.001
Range (mV) ±2000
Resolution (mV) 0.1 mV
Calibration Points 1 to 5 (pH), 1 to 3 (conductivity), 1 (mV)
Temperature Range (Metric) -5°C to +105°C
Resolution (Temperature) 0.1°C
Temperature Probe Calibration 1 point
Display Type LCD, 240 x 64 pixels
Backlight Option White
Data Points 12,000 with Temp, Date and Time Stamp
Inputs 3 BNC for measurement, 3 x 2 banana for temperature
Dimensions (L x W x H) 26W x 18D x 9H cm
Weight (Metric) 1 kg
Warranty 36 Months
Accuracy 0.1% ± 1 digit (rH2)
Accuracy (Conductivity) 0.5% f.s. of range
Model C3050
Range (Conductivity) 0 to 2000 mS/cm
Range (Resistivity) 0 to 200 MΩ.cm
Range (Salinity) 0 - 70
Range (TDS) 0 - 100 g/L
Relative Humidity Range 0 - 95%, non-condensing
Resolution 0.01 rH2 (relative hydrogen), 1 Ω.cm (resistivity)
Resolution (Conductivity) 0.001 μS/cm
Resolution (TDS) 0.01 mg/L
Detection Range rH2: 0 to 42, μW: 0 to 400,000
Temperature Compensation -5°C to +105°C (pH, conductivity)
Temperature (Metric) Operating 0°C to 40°C
Channels 3 measurement (conductivity: 2), 3 temperature
Temperature Sensor Pt1000
Voltage 100/240 VAC (low voltage: 9 - 15 VDC)
Frequency 50/60 Hz