Learn More
Greiner Bio-One 384 Well Polypropylene Flat Bottom Non-Sterile Microplates
Product Code. 10090212
Change view
Click to view available options
Color:
Black
Natural
White
Unit Size:
Pack of 100
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
10090212 Black Pack of 100
10144481 Natural Pack of 100
13047913 White Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10090212 Supplier Greiner Bio-One Supplier No. 781209
Only null left
Add to basket
Add to basket
Resistant to many standard laboratory chemicals such as DMSO. Greiner Bio-One 384 Well Polypropylene Flat Bottom Non-Sterile Microplates are compatible with standard equipment and automated systems.
  • Free of detectable DNase, RNase, human DNA
  • Non-Pyrogenic
  • Manufactured under DIN ISO 9001 guidelinesNumber of wells are quadrupledWell-to-well spacing is 4.5mm
  • Rounded square well profile
  • Non-ste
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Lid Without Lid
Array 16 x 24
Binding Type Non Binding
Color Black
Material Polypropylene
No. of Wells 384
Volume (Metric) Working 15 to 145 μL
Sterility Non-sterile
Total Volume (Metric) Well 152 μL
Well Shape Rounded Square
No. per Case 100
No. per Pack 10
Well Color Black