missing translation for 'onlineSavingsMsg'
Learn More

(3aR,5S,6S,6aR)-5-((R)-2,2-Dimethyl-1,3-dioxolan-4-yl)-2,2-dimethyltetrahydrofuro[2,3-d][1,3]dioxol-6-ol, 97%, BLD Pharm™

Product Code. 31112165
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31112165 10 g
31093501 500 g
31094916 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 31112165 Supplier BLD Pharm Supplier No. BD1425910g
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 582-52-5
Molecular Formula C12H20O6
MDL Number MFCD00005544
Quantity 10 g
SMILES O[C@H]1[C@@H]([C@@H]2OC(C)(C)OC2)O[C@]3([H])OC(C)(C)O[C@@]31[H]
Molecular Weight (g/mol) 260.28
Percent Purity 97%
Chemical Name or Material (3aR,5S,6S,6aR)-5-((R)-2,2-Dimethyl-1,3-dioxolan-4-yl)-2,2-dimethyltetrahydrofuro[2,3-d][1,3]dioxol-6-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.