Learn More
3M™ Fit Test Solution
Product Code. 31605231 Shop All 3M Products
Change view
Click to view available options
Product Type:
Fit Test Kit
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
31605231 Fit Test Kit Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 31605231 Supplier 3M™ Supplier No. 7100350838
  • ONLINE EXCLUSIVE PRICE
    262.00€
    167.10€ / Pack of 6
    Save 94.90€
    36% Off
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Replacement fit test solutions for use with qualitative fit test apparatus kits FT-10 (Sweet) and FT-30 (Bitter).
  • Test kits and accessories help organizations meet regulatory fit testing requirements for tight-fitting respirators.
  • Used to validate the face-to-respirator seal on disposable or half-mask respirators.
  • Support compliance and ensure effective respiratory protection.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Fit Test Kit
With Exhalation Valve No