missing translation for 'onlineSavingsMsg'
Learn More

(3R)-4-Cyano-3-hydroxybutanoic Acid Ethyl Ester, TRC

Product Code. 30369708
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30369708 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30369708 Supplier Toronto Research Chemicals Supplier No. TRCE9059005G
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 141942-85-0
Molecular Formula C7H11NO3
Quantity 5 g
Synonym (3R)-4-Cyano-3-hydroxybutyric Acid Ethyl Ester, (R)-4-Cyano-3-hydroxybutyric Acid Ethyl Ester, (R)-Ethyl 4-Cyano-3-hydroxybutanoate, (R)-Ethyl 4-Cyano-3-hydroxybutanoate, Ethyl (R)-4-Cyano-3-hydroxybutanoate, Ethyl (R)-4-Cyano-3-hydroxybutyrate
InChI Formula InChI=1S/C7H11NO3/c1-2-11-7(10)5-6(9)3-4-8/h6,9H,2-3,5H2,1H3/t6-/m1/s1
SMILES CCOC(=O)C[C@H](O)CC#N
IUPAC Name ethyl (3R)-4-cyano-3-hydroxybutanoate
Molecular Weight (g/mol) 157.17
Formula Weight 157.0739
Chemical Name or Material (3R)-4-Cyano-3-hydroxybutanoic Acid Ethyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.