missing translation for 'onlineSavingsMsg'
Learn More

(3RS)-3-(4-Chloro-phenyl)-N-methyl-3-(pyridin-2-yl)propan-1-amine Maleate, Mikromol™

Product Code. 18063930
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18063930 100 mg 100mg
18020540 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18063930 Supplier Mikromol Supplier No. MM0090.13
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(3RS)-3-(4-Chloro-phenyl)-N-methyl-3-(pyridin-2-yl)propan-1-amine Maleate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Desmethylchlorphenamine Maleate
CAS 22630-25-7
Analyte or Component Names Desmethylchlorphenamine Maleate
Shipping Condition Room Temperature
Molecular Formula C15 H17 Cl N2 . C4 H4 O4
Certifications/Compliance ISO 17025
Synonym (3RS)-3-(4-Chloro-phenyl)-N-methyl-3-(pyridin-2-yl)propan-1-amine Maleate, Chlorphenamine Maleate Imp. C (EP) as Maleate, Desmethylchlorphenamine Maleate, Chlorphenamine Maleate Imp. C (EP), Chlorphenamine Imp. C (EP), Chlorpheniramine USP Related Compound C, Chlorpheniramine USP RC C, Chlorpheniramine Related Compound C as Maleate, Chlorphenamine Maleate Impurity C as Maleate, Chlorphenamine Impurity C as Maleate
InChI Formula InChI=1S/C15H17ClN2.C4H4O4/c1-17-11-9-14(15-4-2-3-10-18-15)12-5-7-13(16)8-6-12;5-3(6)1-2-4(7)8/h2-8,10,14,17H,9,11H2,1H3;1-2H,(H,5,6)(H,7,8)/b;2-1-
SMILES CNCCC(c1ccc(Cl)cc1)c2ccccn2.OC(=O)\C=C/C(=O)O
IUPAC Name (Z)-but-2-enedioic acid;3-(4-chlorophenyl)-N-methyl-3-pyridin-2-ylpropan-1-amine
Molecular Weight (g/mol) 376.83
Quantity 100 mg
Formula Weight 376.119 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.