missing translation for 'onlineSavingsMsg'
Learn More

(3RS)-4-(Dimethylamino)-3-methyl-2,2-diphenylbutanenitrile (Isodidiavalo), Mikromol™

Product Code. 18062480
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18062480 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18062480 Supplier Mikromol Supplier No. MM0531.12
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(3RS)-4-(Dimethylamino)-3-methyl-2,2-diphenylbutanenitrile (Isodidiavalo)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Isodidiavalo
CAS 1-2-6293
Analyte or Component Names Isodidiavalo
Shipping Condition Room Temperature
Molecular Formula C19 H22 N2
Certifications/Compliance ISO 17025
Synonym Methadone Hydrochoride Imp. C (EP), (3RS)-4-(Dimethylamino)-3-methyl-2,2-diphenylbutanenitrile, Isodidiavalo, alpha-[2-(Dimethylamino)-1-methylethyl]-alpha-phenylbenzeneacetonitrile, 4-(Dimethylamino)-3-methyl-2,2-diphenylbutyronitrile, 2,2-Diphenyl-3-methyl-4-(dimethylamino)butyronitrile, NSC 9993, Tuso-Blandin, Methadone Imp. C (EP), Methadone Related Compound C, Methadone Hydrochloride Impurity C, Methadone Impurity C
InChI Formula InChI=1S/C19H22N2/c1-16(14-21(2)3)19(15-20,17-10-6-4-7-11-17)18-12-8-5-9-13-18/h4-13,16H,14H2,1-3H3
SMILES CC(CN(C)C)C(C#N)(c1ccccc1)c2ccccc2
IUPAC Name 4-(dimethylamino)-3-methyl-2,2-diphenylbutanenitrile
Molecular Weight (g/mol) 278.39
Quantity 50 mg
Formula Weight 278.1783 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.