missing translation for 'onlineSavingsMsg'
Learn More

(3RS)-5-Amino-3-(4-chlorophenyl)-5-oxopentanoic Acid, Mikromol™

Product Code. 30050880
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30050880 100 mg 100mg
30049856 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30050880 Supplier Mikromol Supplier No. MM1235.03
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(3RS)-5-Amino-3-(4-chlorophenyl)-5-oxopentanoic Acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (3RS)-5-Amino-3-(4-chlorophenyl)-5-oxopentanoic Acid
CAS 1141-23-7
Analyte or Component Names (3RS)-5-Amino-3-(4-chlorophenyl)-5-oxopentanoic Acid
Shipping Condition Room Temperature
Molecular Formula C11 H12 Cl N O3
Certifications/Compliance ISO 17025
Synonym (3RS)-5-Amino-3-(4-chlorophenyl)-5-oxopentanoic Acid, Baclofen Imp. B (EP), Baclofen Impurity B
InChI Formula InChI=1S/C11H12ClNO3/c12-9-3-1-7(2-4-9)8(5-10(13)14)6-11(15)16/h1-4,8H,5-6H2,(H2,13,14)(H,15,16)
SMILES NC(=O)CC(CC(=O)O)c1ccc(Cl)cc1
IUPAC Name 5-amino-3-(4-chlorophenyl)-5-oxopentanoic acid
Molecular Weight (g/mol) 241.67
Quantity 100 mg
Formula Weight 241.0506 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.