missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(3RS)-N-Methyl-3-phenyl-3-[3-(trifluoromethyl)phenoxy]propan-1-amine Hydrochloride, Mikromol™
Description
(3RS)-N-Methyl-3-phenyl-3-[3-(trifluoromethyl)phenoxy]propan-1-amine Hydrochloride
Specifications
Specifications
| Chemical Name or Material | (3RS)-N-Methyl-3-phenyl-3-[3-(trifluoromethyl)phenoxy]propan-1-amine Hydrochloride |
| CAS | 79088-29-2 |
| Analyte or Component Names | (3RS)-N-Methyl-3-phenyl-3-[3-(trifluoromethyl)phenoxy]propan-1-amine Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H18 F3 N O . Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | Fluoxetine Hydrochloride Imp. C (EP), Fluoxetine Imp. C (EP), Fluoxetine USP Related Compound A, Fluoxetine USP RC A, (3RS)-N-Methyl-3-phenyl-3-[3-(trifluoromethyl)phenoxy]propan-1-amine Hydrochloride, Fluoxetine Hydrochloride Imp. C (EP) as Hydrochloride, Fluoxetine Related Compound A as Hydrochloride, Fluoxetine Hydrochloride Impurity C as Hydrochloride, Fluoxetine Impurity C as Hydrochloride |
| InChI Formula | InChI=1S/C17H18F3NO.ClH/c1-21-11-10-16(13-6-3-2-4-7-13)22-15-9-5-8-14(12-15)17(18,19)20;/h2-9,12,16,21H,10-11H2,1H3;1H |
| SMILES | Cl.CNCCC(Oc1cccc(c1)C(F)(F)F)c2ccccc2 |
| IUPAC Name | N-methyl-3-phenyl-3-[3-(trifluoromethyl)phenoxy]propan-1-amine;hydrochloride |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?