missing translation for 'onlineSavingsMsg'
Learn More

(3S,5S)-Atorvastatin Sodium Salt, TRC

Product Code. 30011497
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011497 25 mg 25mg
30006385 10 mg 10mg
30007025 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011497 Supplier Toronto Research Chemicals Supplier No. TRCA79173525MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1428118-38-0
Molecular Formula C33 H34 F N2 O5 . Na
Molecular Weight (g/mol) 580.62
Synonym 1H-Pyrrole-1-heptanoic acid, 2-(4-fluorophenyl)-β,δ-dihydroxy-5-(1-methylethyl)-3-phenyl-4-[(phenylamino)carbonyl]-, sodium salt (1:1), (βS,δS)-, (3S,5S)-7-[2-(4-Fluorophenyl)-5-(1-methylethyl)-3-phenyl-4-(phenylcarbamoyl)-1H-pyrrol-1-yl]-3,5-dihydroxyheptanoic acid sodium salt, (βS,δS)-2-(4-Fluorophenyl)-β,δ-dihydroxy-5-(1-methylethyl)-3-phenyl-4-[(phenylamino)carbonyl]-1H-pyrrole-1-heptanoic acid sodium salt (1:1), (3S,5S)-Atorvastatin sodium salt, ent-Atorvastatin sodium salt, Atorvastatin Imp. E (EP) as Sodium Salt
IUPAC Name sodium;(3S,5S)-7-[2-(4-fluorophenyl)-3-phenyl-4-(phenylcarbamoyl)-5-propan-2-ylpyrrol-1-yl]-3,5-dihydroxyheptanoate
SMILES [Na+].CC(C)c1c(C(=O)Nc2ccccc2)c(c3ccccc3)c(c4ccc(F)cc4)n1CC[C@H](O)C[C@H](O)CC(=O)[O-]

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C33H35FN2O5.Na/c1-21(2)31-30(33(41)35-25-11-7-4-8-12-25)29(22-9-5-3-6-10-22)32(23-13-15-24(34)16-14-23)36(31)18-17-26(37)19-27(38)20-28(39)40;/h3-16,21,26-27,37-38H,17-20H2,1-2H3,(H,35,41)(H,39,40);/q;+1/p-1/t26-,27-;/m0./s1
Formula Weight 580.2349
Chemical Name or Material ent-Atorvastatin sodium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.