missing translation for 'onlineSavingsMsg'
Learn More

[4-(1,3-Thiazol-2-yl)phenyl]methanol, 97%, Thermo Scientific™

Product Code. 10554974
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10554974 5 g Amber Glass Bottle 5g
10453555 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10554974 Supplier Thermo Scientific Maybridge Supplier No. CC40209DE
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 454678-91-2
Molecular Formula C10H9NOS
Molecular Weight (g/mol) 191.248
MDL Number MFCD09064950
InChI Key QOLUYHARONOGNO-UHFFFAOYSA-N
Synonym 4-1,3-thiazol-2-yl phenyl methanol, benzenemethanol,4-2-thiazolyl, 4-thiazol-2-yl benzyl alcohol, 4-thiazol-2-yl phenyl methanol, 4-1,3-thiazol-2-yl benzyl alcohol, 4-thiazol-2-yl benzylalcohol, 4-1,3-thiazol-2-yl phenyl methan-1-ol
PubChem CID 24229487
IUPAC Name [4-(1,3-thiazol-2-yl)phenyl]methanol
SMILES C1=CC(=CC=C1CO)C2=NC=CS2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 191.25
Percent Purity 97%
Chemical Name or Material [4-(1, 3-Thiazol-2-yl)phenyl]methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.