missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-[1-[4-(Aminosulfonyl)phenyl]-3-(trifluoromethyl)-1H-pyrazol-5-yl]benzoic Acid, Mikromol™
Description
4-[1-[4-(Aminosulfonyl)phenyl]-3-(trifluoromethyl)-1H-pyrazol-5-yl]benzoic Acid
Specifications
Specifications
| Chemical Name or Material | Celecoxib Carboxylic Acid |
| CAS | 170571-01-4 |
| Analyte or Component Names | Celecoxib Carboxylic Acid |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H12 F3 N3 O4 S |
| Certifications/Compliance | ISO 17025 |
| Synonym | 4-[1-[4-(Aminosulfonyl)phenyl]-3-(trifluoromethyl)-1H-pyrazol-5-yl]benzoic Acid, Celecoxib Carboxylic Acid |
| InChI Formula | InChI=1S/C17H12F3N3O4S/c18-17(19,20)15-9-14(10-1-3-11(4-2-10)16(24)25)23(22-15)12-5-7-13(8-6-12)28(21,26)27/h1-9H,(H,24,25)(H2,21,26,27) |
| SMILES | NS(=O)(=O)c1ccc(cc1)n2nc(cc2c3ccc(cc3)C(=O)O)C(F)(F)F |
| IUPAC Name | 4-[2-(4-sulfamoylphenyl)-5-(trifluoromethyl)pyrazol-3-yl]benzoic acid |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?