missing translation for 'onlineSavingsMsg'
Learn More

4-[1-[4-(Aminosulfonyl)phenyl]-3-(trifluoromethyl)-1H-pyrazol-5-yl]benzoic Acid, Mikromol™

Product Code. 30460442
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30460442 4 x 25 mg 100mg
30049801 25 mg 25mg
17931006 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30460442 Supplier Mikromol Supplier No. MM0344.0300254AMP
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

4-[1-[4-(Aminosulfonyl)phenyl]-3-(trifluoromethyl)-1H-pyrazol-5-yl]benzoic Acid, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Celecoxib Carboxylic Acid
CAS 170571-01-4
Analyte or Component Names Celecoxib Carboxylic Acid
Shipping Condition Room Temperature
Molecular Formula C17 H12 F3 N3 O4 S
Certifications/Compliance ISO 17025
Synonym 4-[1-[4-(Aminosulfonyl)phenyl]-3-(trifluoromethyl)-1H-pyrazol-5-yl]benzoic Acid, Celecoxib Carboxylic Acid
InChI Formula InChI=1S/C17H12F3N3O4S/c18-17(19,20)15-9-14(10-1-3-11(4-2-10)16(24)25)23(22-15)12-5-7-13(8-6-12)28(21,26)27/h1-9H,(H,24,25)(H2,21,26,27)
Solution Type Neat
SMILES NS(=O)(=O)c1ccc(cc1)n2nc(cc2c3ccc(cc3)C(=O)O)C(F)(F)F
IUPAC Name 4-[2-(4-sulfamoylphenyl)-5-(trifluoromethyl)pyrazol-3-yl]benzoic acid
Molecular Weight (g/mol) 411.36
Quantity 4 x 25 mg
Formula Weight 411.0501 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.