Learn More
4-(1-Adamantyl)phenol, TRC
Specifications
Specifications
| Chemical Name or Material | 4-(1-Adamantyl)phenol |
| CAS | 29799-07-3 |
| Molecular Formula | C16H20O |
| Quantity | 5 g |
| Synonym | p-1-Adamantyl-phenol, 1-(4-Hydroxyphenyl)adamantane, 1-(p-Hydroxyphenyl)adamantane, 4-(Hydroxyphenyl)adamantane, NSC 111653, p-1-Adamantylphenol, 4-Tricyclo[3.3.1.13,7]dec-1-yl-phenol |
| InChI Formula | InChI=1S/C16H20O/c17-15-3-1-14(2-4-15)16-8-11-5-12(9-16)7-13(6-11)10-16/h1-4,11-13,17H,5-10H2 |
| SMILES | OC(=O)[C@@H](N)Cc1ncnc1 |
| IUPAC Name | 4-(1-adamantyl)phenol |
| Molecular Weight (g/mol) | 228.33 |
| Formula Weight | 228.1514 |