missing translation for 'onlineSavingsMsg'
Learn More

4-(1-Pyrrolidinylmethyl)benzeneboronic acid pinacol ester, Thermo Scientific™

Product Code. 15459155
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15459155 1 g 1g
15469155 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15459155 Supplier Thermo Scientific Alfa Aesar Supplier No. H51115.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 884507-39-5
Molecular Formula C17H26BNO2
Molecular Weight (g/mol) 287.21
MDL Number MFCD08690299
InChI Key AGSIDMRVRGPBIE-UHFFFAOYSA-N
Synonym 1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzyl pyrrolidine, 1-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl pyrrolidine, 1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl pyrrolidine, 4-pyrrolidine methyl phenylboronic acid pinacol ester, pyrrolidine,1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl, 4-pyrrolidinomethyl phenylboronic acid, pinacol ester, amtb121, 4-1-pyrroldinylmethyl benzeneboronic acid pinacol ester, 4-pyrrolidin-1-ylmethyl phenylboronic acid pinacol ester
PubChem CID 18525868
IUPAC Name 1-[[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl]pyrrolidine
SMILES CC1(C)OB(OC1(C)C)C1=CC=C(CN2CCCC2)C=C1

Specifications

Quantity 1 g
Formula Weight 287.2
Chemical Name or Material 4-(1-Pyrrolidinylmethyl)benzeneboronic acid pinacol ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.