missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-1BB Ligand/TNFSF9 Antibody (AP-MAB0875) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-06697-0.1mg
This item is not returnable.
View return policy
Description
4-1BB Ligand/TNFSF9 Monoclonal specifically detects 4-1BB Ligand/TNFSF9 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen, CyTOF-ready, Block/Neutralize.
Specifications
| 4-1BB Ligand/TNFSF9 | |
| Monoclonal | |
| 1 mg/mL | |
| Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Blocking/Neutralizing 50-200 molar excess, CyTOF-ready | |
| P41274 | |
| TNFSF9 | |
| BALB/c mouse B lymphoma cell line | |
| 0.1 mg | |
| Apoptosis | |
| 8744 | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| AP-MAB0875 | |
| Unconjugated | |
| PBS with No Preservative | |
| 4-1BB ligand, 4-1BBL, 4-1BB-L, CD137L, homolog of mouse 4-1BB-L, receptor 4-1BB ligand, tumor necrosis factor (ligand) superfamily, member 9, tumor necrosis factor ligand superfamily member 9 | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| This was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat) immunized with BALB/c mouse B lymphoma cell line. | |
| Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction