Learn More
4-(1H-Pyrazol-1-ylmethyl)aniline, 95%, Thermo Scientific™
Product Code. 10420754
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10420754 250 mg 250mg
10476853 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10420754 Supplier Thermo Scientific Maybridge Supplier No. CC56014CB
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Chemical Identifiers
CAS 142335-61-3
Molecular Formula C10H11N3
Molecular Weight (g/mol) 173.22
MDL Number MFCD03422514
InChI Key ISRYTHBRUSOVAB-UHFFFAOYSA-N
Synonym 4-1h-pyrazol-1-ylmethyl aniline, 4-1h-pyrazol-1-yl methyl aniline, 4-pyrazol-1-ylmethyl aniline, 4-1-pyrazolyl methyl aniline, 4-pyrazol-1-ylmethyl-phenylamine, 4-pyrazol-1-ylmethylphenylamine, benzenamine,4-1h-pyrazol-1-ylmethyl, 4-pyrazolylmethyl phenylamine, pyrazolylmethylaniline
PubChem CID 2764412
IUPAC Name 4-(pyrazol-1-ylmethyl)aniline
SMILES NC1=CC=C(CN2C=CC=N2)C=C1
Specifications
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Quantity 250 mg
Formula Weight 173.22
Percent Purity 95%
Chemical Name or Material 4-(1H-Pyrazol-1-ylmethyl)aniline