missing translation for 'onlineSavingsMsg'
Learn More

[4-[2-[[(3-Ethyl-2,5-dihydro-4-methyl-2-oxo-1H-pyrrol-1-yl)carbonyl]amino]ethyl]phenyl]sulfonyl]carbamic Acid Ethyl Ester, TRC

Product Code. 17932785
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17932785 100 mg 100mg
17991475 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17932785 Supplier Toronto Research Chemicals Supplier No. TRCE916540100MG
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 318515-70-7
Molecular Weight (g/mol) 423.48
SMILES CCOC(=O)NS(=O)(=O)c1ccc(CCNC(=O)N2CC(=C(CC)C2=O)C)cc1

Specifications

Molecular Formula C19H25N3O6S
Quantity 100 mg
Synonym N-[[4-[2-[[(3-Ethyl-2,5-dihydro-4-methyl-2-oxo-1H-pyrrol-1-yl)carbonyl]amino]ethyl]phenyl]sulfonyl]carbamic Acid Ethyl Ester, 4-[2-[(3-Ethyl-4-methyl-2-oxo-3-pyrrolin-1-yl)carboxamido]ethyl]phenyl]sulfonyl]carbamic Acid Ethyl Ester
InChI Formula InChI=1S/C19H25N3O6S/c1-4-16-13(3)12-22(17(16)23)18(24)20-11-10-14-6-8-15(9-7-14)29(26,27)21-19(25)28-5-2/h6-9H,4-5,10-12H2,1-3H3,(H,20,24)(H,21,25)
Formula Weight 423.1464
Percent Purity >95%
Chemical Name or Material Ethyl ([4-[2-([(3-Ethyl-4-methyl-2-oxo-2,5-dihydro-1H-pyrrol-1-yl)carbonyl]amino)ethyl]phenyl]sulphonyl)carbamate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.