missing translation for 'onlineSavingsMsg'
Learn More

4-[2-(5-Chloro-2-methoxybenzamido)ethyl]benzene Sulfonamide, TRC

Product Code. 30006953
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30006953 5 g 5g
30003495 10 g 10g
30006716 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30006953 Supplier Toronto Research Chemicals Supplier No. TRCC3678955G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16673-34-0
Molecular Formula C16 H17 Cl N2 O4 S
Molecular Weight (g/mol) 368.84
Synonym 5-Chloro-2-methoxy-N-[2-(4-sulfamoylphenyl)ethyl]benzamide, Glibenclamide Imp. A (EP)
IUPAC Name 5-chloro-2-methoxy-N-[2-(4-sulfamoylphenyl)ethyl]benzamide
SMILES COc1ccc(Cl)cc1C(=O)NCCc2ccc(cc2)S(=O)(=O)N

Specifications

Quantity 5 g
InChI Formula InChI=1S/C16H17ClN2O4S/c1-23-15-7-4-12(17)10-14(15)16(20)19-9-8-11-2-5-13(6-3-11)24(18,21)22/h2-7,10H,8-9H2,1H3,(H,19,20)(H2,18,21,22)
Formula Weight 368.0598
Percent Purity >95%
Chemical Name or Material 5-Chloro-2-methoxy-N-[2-(4-sulfamoylphenyl)ethyl]benzamide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.