Learn More
4'-(2-Chloroethyl)acetophenone, tech. 90%, Thermo Scientific™
Product Code. 11383257
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. 11383257
Brand: Thermo Scientific Alfa Aesar L13800.06
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 69614-95-5
Molecular Formula C10H11ClO
Molecular Weight (g/mol) 182.647
MDL Number MFCD00016350
InChI Key RGVUACHOYCYNMB-UHFFFAOYSA-N
Synonym 4'-beta-chloroethyl acetophenone, 4-2-chloroethyl acetophenone, 1-4-2-chloroethyl phenyl ethanone, 4'-2-chloroethyl acetophenone, 1-4-2-chloroethyl phenyl-1-ethanone, 4'-.beta.-chloroethyl acetophenone, p-beta-chloroethyl acetophenone, 1-4-2-chloroethyl phenyl ethan-1-one, 1-acetyl-4-2-chloroethyl benzene
PubChem CID 144359
IUPAC Name 1-[4-(2-chloroethyl)phenyl]ethanone
SMILES CC(=O)C1=CC=C(C=C1)CCCl
Specifications
Density 1.15
Boiling Point 112°C (0.1 mmHg)
Refractive Index 1.555
Quantity 5 g
Beilstein 2081019
Formula Weight 182.65
Percent Purity 90%
Grade Technical
Chemical Name or Material 4'-(2-Chloroethyl)acetophenone