missing translation for 'onlineSavingsMsg'
Learn More

4'-(2-Chloroethyl)acetophenone, tech. 90%, Thermo Scientific™

Product Code. 11393257
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11393257

Brand: Thermo Scientific Alfa Aesar L13800.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69614-95-5
Molecular Formula C10H11ClO
Molecular Weight (g/mol) 182.647
MDL Number MFCD00016350
InChI Key RGVUACHOYCYNMB-UHFFFAOYSA-N
Synonym 4'-beta-chloroethyl acetophenone, 4-2-chloroethyl acetophenone, 1-4-2-chloroethyl phenyl ethanone, 4'-2-chloroethyl acetophenone, 1-4-2-chloroethyl phenyl-1-ethanone, 4'-.beta.-chloroethyl acetophenone, p-beta-chloroethyl acetophenone, 1-4-2-chloroethyl phenyl ethan-1-one, 1-acetyl-4-2-chloroethyl benzene
PubChem CID 144359
IUPAC Name 1-[4-(2-chloroethyl)phenyl]ethanone
SMILES CC(=O)C1=CC=C(C=C1)CCCl

Specifications

Density 1.15
Boiling Point 112°C (0.1 mmHg)
Refractive Index 1.555
Quantity 25 g
Beilstein 2081019
Formula Weight 182.65
Percent Purity 90%
Grade Technical
Chemical Name or Material 4'-(2-Chloroethyl)acetophenone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.