missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Chloroethyl)morpholine Hydrochloride, TRC

Product Code. 30008291
Change view
Click to view available options
Quantity:
10 g
100 g
Unit Size:
100g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30008291 100 g 100g
30003439 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30008291 Supplier Toronto Research Chemicals Supplier No. TRCC366195100G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3647-69-6
Molecular Formula C6H12ClNO . HCl
Molecular Weight (g/mol) 186.08
Synonym 1-(Morpholin-4-yl)-2-chloroethane Hydrochloride, 1-Chloro-2-(morpholin-4-yl)ethane Monohydrochloride, 1-Chloro-2-morpholinoethane Hydrochloride, 2-(4-Morpholinyl)ethyl Chloride Hydrochloride, 2-Morpholinoethyl Chloride Hydrochloride, 4-(2-Chloroethyl)morpholine Monohydrochloride, 4-(2-Chloroethyl)morpholinium Chloride, Morpholinoethyl chloride Hydrochloride, N-(2-Chloroethyl)morpholine Hydrochloride, N-(β-Chloroethyl)morpholine Hydrochloride, β-Morpholinoethyl Chloride Hydrochloride;
IUPAC Name 4-(2-chloroethyl)morpholine;hydrochloride
SMILES Cl.ClCCN1CCOCC1

Specifications

Quantity 100 g
InChI Formula InChI=1S/C6H12ClNO.ClH/c7-1-2-8-3-5-9-6-4-8;/h1-6H2;1H
Chemical Name or Material 4-(2-Chloroethyl)morpholine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.