missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Chloronicotinoyl)morpholine, 97%, Thermo Scientific™

Product Code. 11325619
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11325619 1 g 1g
11315619 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11325619 Supplier Thermo Scientific Alfa Aesar Supplier No. H50522.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53062-98-9
Molecular Formula C10H11ClN2O2
Molecular Weight (g/mol) 226.66
MDL Number MFCD00454210
InChI Key SPSWDQBZCCVHNI-UHFFFAOYSA-N
Synonym 4-2-chloronicotinoyl morpholine, 4-2-chloropyridine-3-carbonyl morpholine, 4-2-chloropyridin-3-yl carbonyl morpholine, 2-chloro-3-pyridinyl 4-morpholinyl methanone, 2-chloropyridin-3-yl morpholino methanone, 4-2-chloro-3-pyridinyl carbonyl morpholine, acmc-20amr1, 2-chloronicotinic acid morpholide, 2-chloro 3-pyridyl morpholin-4-yl ketone, 2-chloro-3-pyridinyl morpholino methanone
PubChem CID 2063428
SMILES ClC1=C(C=CC=N1)C(=O)N1CCOCC1

Specifications

Quantity 1 g
IUPAC Name (2-chloropyridin-3-yl)-morpholin-4-ylmethanone
Formula Weight 226.66
Percent Purity 97%
Chemical Name or Material 4-(2-Chloronicotinoyl)morpholine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.