missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Ethyl-6-methylphenyl)-2-hydroxy-5-methyl-3-morpholinone, TRC

Product Code. 30180394
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30180394 500 mg 500mg
30180169 1 g 1g
30179737 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30180394 Supplier Toronto Research Chemicals Supplier No. TRCE904560500MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61520-54-5
Molecular Formula C14 H19 N O3
Molecular Weight (g/mol) 249.3056
Synonym 3-Morpholinone, 4-(2-ethyl-6-methylphenyl)-2-hydroxy-5-methyl-, 4-(2-Ethyl-6-methylphenyl)-2-hydroxy-5-methyl-3-morpholinone, CGA 49751
IUPAC Name 4-(2-ethyl-6-methylphenyl)-2-hydroxy-5-methylmorpholin-3-one
SMILES CCc1cccc(C)c1N2C(C)COC(O)C2=O

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C14H19NO3/c1-4-11-7-5-6-9(2)12(11)15-10(3)8-18-14(17)13(15)16/h5-7,10,14,17H,4,8H2,1-3H3
Formula Weight 249.1365
Chemical Name or Material Metolachlor metabolite CGA 49751
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.